Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calcium dihydrogen phosphate hydrate, 97%

Catalog No. p-7081864
Change view
Click to view available options
Quantity:
10,000 g
2500 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1212336 500 g
AAA121230E 2500 g
AAA121230C 10,000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1212336 Supplier Thermo Scientific Chemicals Supplier No. A1212336
Only null left
Add to Cart

CAS: 301524-28-7 | CaH4O8P2 | 234.05 g/mol

Calcium dihydrogen phosphate hydrate is a pH control reagent, stabilizer for plastics, manufacture of glass.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Calcium dihydrogen phosphate hydrate is a pH control reagent, stabilizer for plastics, manufacture of glass.

Solubility
Soluble in water 18 g/L @ 30°C.

Notes
Loses H{2}O at 100°, dec. at 200°
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 301524-28-7
Molecular Formula CaH4O8P2
Molecular Weight (g/mol) 234.05
MDL Number MFCD00010898
InChI Key YYRMJZQKEFZXMX-UHFFFAOYSA-L
Synonym Calcium phosphate, monobasic; Calcium tetrahydrogen diphosphate
IUPAC Name calcium didihydrogen phosphate
SMILES [Ca++].OP(O)([O-])=O.OP(O)([O-])=O

Specifications

Melting Point ∼200°C (decomposition)
Quantity 500 g
Flash Point None
Merck Index 14,1693
Solubility Information Soluble in water 18g/L @ 30°C.
Formula Weight 234.06 (Anhydrous)
Percent Purity 97%
Chemical Name or Material Calcium dihydrogen phosphate hydrate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 231-837-1
RTECSNumber TB8527000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.