Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calcium Disodium EDTA, FCC, 97-102%, Spectrum™ Chemical
SDP

Catalog No. 18601295 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
125 g
500 g
2.5 kg
12 kg
45 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-601-295 12 kg Poly Pail
18-601-291 125 g Poly Bottle
18-601-292 500 g Poly Bottle
18-601-293 2.5 kg Poly Pail
18-601-294 45 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18-601-295 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. E119412KG

Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.

E1194, 23411-34-9, C10H12CaN2Na2O8·xH2O

Spectrum™ Chemical Calcium Disodium EDTA, FCC is used as a flavor and color preservative. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 23411-34-9
Molecular Formula C10H16CaN2Na2O9
Molecular Weight (g/mol) 394.30
InChI Key WJEKUTCBXSHIOX-UHFFFAOYSA-J
SMILES O.[Na+].[Na+].[Ca+4].[O-]C(=O)C[NH](CC[NH](CC([O-])=O)CC([O-])=O)CC([O-])=O

Specifications

pH 6.5-7.5
CAS Max % 100%
Quantity 12 kg
Percent Purity 97 to 102%
Grade FCC
Packaging Poly Pail
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.