Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calcium fluoride, ultra dry, 99.99% (metals basis)

Catalog No. AA3574222
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3574222 100 g
AA3574206 5 g
AA3574214 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA3574222 Supplier Thermo Scientific Chemicals Supplier No. 03574222
 Restricted Item
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 7789-75-5 | CaF2 | 78.08 g/mol

Calcium fluoride is the commercially important source for fluoride compounds as well as hydrogen fluoride. It is transparent from ultraviolet to infrared region. It is used in spectroscopy, thermal imaging systems and excimer lasers; it is also used in the manufacturing of optical components including windows, prisms and lenses. In addition, it is also used in aluminum-metallurgy, brake lining manufacturing industries, and in tooth enamel.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7789-75-5
Molecular Formula CaF2
Molecular Weight (g/mol) 78.08
MDL Number MFCD00010907
InChI Key WUKWITHWXAAZEY-UHFFFAOYSA-L
Synonym calcium fluoride, kalziumfluorid, kalziumdifluorid, calcium 2+ difluoride, acmc-1bk0c, ksc377c5h, calcium fluoride 100g, calcium fluoride,optical grade
PubChem CID 84512
ChEBI CHEBI:35437
SMILES [F-].[F-].[Ca++]

Specifications

Boiling Point ca 2500°C
Melting Point 1403°C
Color White
Physical Form Powder
Quantity 100 g
Assay Percent Range 99.99% (Metals basis)
Beilstein EM>5,
Solubility Information Slightly soluble in acid. Insoluble in acetone.
IUPAC Name calcium;difluoride
Formula Weight 78.08
Odor Odorless
Refractive Index 1.434
Sensitivity Hygroscopic
Density 3.18
Chemical Name or Material Calcium fluoride
Show More Show Less
Hazard Category H315-H319-H335-H303 : Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. May be harmful if swallowed.
EINECSNumber 232-188-7
RTECSNumber EW1760000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.