Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Calcium Flux Assay Kit, Fura 2, Ratiometric, 100x96 Tests

Catalog No. 502582276
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 x 96 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-258-2276 100 x 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-258-2276 Supplier Abcam Supplier No. AB176766100X96TESTS
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Request a Quote

Measure calcium flux in cell samples in 1 hr 20 min using a fluorometric plate reader.

  • Assay Type: Cell-based
  • Detection Method: Fluorescent
  • Assay Time: 1 hr 20 min
  • Sample Types: Suspension cells, Adherent cells

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Target Molecule Calcium
Substrate Properties Chemical Substrate
Substrate Fura-2
Label or Dye Fura-2
No. of Reactions 9600 Tests
For Use With (Equipment) Fluorometric Plate Reader
Detection Location In-Cell Detection
Format Kit
Detection Method Fluorescence
Sample Type Suspension cells, Adherent cells
Quantity 100 x 96 Tests
For Use With (Application) Calcium flux measurement
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.