Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calcium glycerophosphate hydrate, 97%

Catalog No. AAA162690B
Change view
Click to view available options
Quantity:
250 g
1000 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA162690B 1000 g
AAA1626930 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA162690B Supplier Thermo Scientific Chemicals Supplier No. A162690B
Add to Cart

CAS: 28917-82-0 | C3H7CaO6P | 210.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 28917-82-0
Molecular Formula C3H7CaO6P
Molecular Weight (g/mol) 210.14
MDL Number MFCD00042643
InChI Key IWIRHXNCFWGFJE-UHFFFAOYNA-L
Synonym calcium glycerophosphate hydrate, calcii glycerophosphas, c3h7o6p.ca.h2o, glycerophosphoric acid calcium salt hydrate, calcium 2,3-dihydroxypropyl phosphate hydrate, calcium dl-glycerol 3-phosphate hydrate
PubChem CID 71311563
IUPAC Name calcium 3-(phosphonatooxy)propane-1,2-diol
SMILES [Ca++].OCC(O)COP([O-])([O-])=O

Specifications

Quantity 1000 g
Merck Index 14,1670
Formula Weight 210.15 (Anhydrous)
Percent Purity 97%
Chemical Name or Material Calcium glycerophosphate hydrate
EINECSNumber 249-312-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.