Learn More
Calcium hydride, coarse powder, ca 92%
Description
Calcium hydride is used as an efficient drying agent for aprotic base-stable solvents like ethers and tertiary amines. It is a useful dehydrating agent in a synthesis of aldehyde enamines in high yield and purity.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Calcium chloride is used as an efficient drying agent for aprotic base-stable solvents like ethers and tertiary amines. It is a useful dehydrating agent in a synthesis of aldehyde enamines in high yield and purity.
Solubility
Soluble in water and alcohol. Insoluble in benzene.
Notes
Moisture sensitive. Reacts with water vigorously and liberates hydrogen gas. Incompatible with strong oxidizing agents, strong acids, halogens, water and alcohols.
Specifications
Specifications
| Melting Point | 600°C (decomposition) |
| Physical Form | Coarse Powder |
| Quantity | 1000 g |
| UN Number | UN1404 |
| Merck Index | 14,1672 |
| Solubility Information | Soluble in water and alcohol. Insoluble in benzene. |
| Formula Weight | 42.1 |
| Percent Purity | ∼92% |
| Odor | Odorless |
| Density | 1.9 |
| Show More |
Safety and Handling
| Hazard Category | H260-H315-H319 |
| Hazard Statement | GHS H Statement H260 In contact with water releases flammable gases which may ignite spontaneously. |
| Precautionary Statement | P223-P231+P232-P264b-P280-P302+P352-P305+P351+P338-P332+P313-P335+P334-P362-P370+P378q-P501c |
| DOTInformation | Transport Hazard Class: 4.3; Packing Group: I; Proper Shipping Name: CALCIUM HYDRIDE |
| EINECSNumber | 232-189-2 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.