Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calcium hydroxide, 98%, extra pure

Catalog No. p-3734701
Change view
Click to view available options
Quantity:
100 g
500 g
1 kg
2.5 kg
10 kg
Packaging:
Plastic bottle
Plastic drum
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC219181000 100 g Plastic bottle
AC219185000 500 g Plastic bottle
AC219180010 1 kg Plastic bottle
AC219180025 2.5 kg Plastic bottle
AC219180100 10 kg Plastic drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. AC219181000 Supplier Thermo Scientific Chemicals Supplier No. 219181000
Only null left
Add to Cart

CAS: 1305-62-0 | CaH2O2 | 74.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1305-62-0
Molecular Formula CaH2O2
Molecular Weight (g/mol) 74.09
MDL Number MFCD00010901
InChI Key AXCZMVOFGPJBDE-UHFFFAOYSA-L
Synonym calcium hydroxide, slaked lime, hydrated lime, calcium dihydroxide, calcium ii hydroxide, calcium oxide, hydrated, kalziumhydroxid, loeschkalk, pickling lime, calciumdihydroxid
PubChem CID 6093208
ChEBI CHEBI:31341
SMILES [OH-].[OH-].[Ca++]

Specifications

Boiling Point 2850.0°C
Melting Point 580.0°C
Color White
Physical Form Amorphous Powder
Quantity 100 g
Assay Percent Range 97.5% min. (Complexometry)
Linear Formula Ca(OH)2
Merck Index 15, 1675
Solubility Information Solubility in water: 1.65g/L (20°C). Other solubilities: soluble in acids (evolution of heat) and nh4cl sol, soluble in glycerol and sugar, insoluble in alcohol
IUPAC Name calcium;dihydroxide
Formula Weight 74.09
Percent Purity 98%
Grade Extra Pure
Identification Passes Tests
Packaging Plastic bottle
Density 2.24g/mL
Chemical Name or Material Calcium hydroxide
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes serious eye damage.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately
EINECSNumber 215-137-3

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.