Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calcium Hydroxide, Reag. Ph. Eur., puriss. p.a., Solstice

Catalog No. 60045923 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
1 kg
25 kg
6 x 1 kg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-045-923 6 x 1 kg
60-023-18 100 g
60-023-21 500 g
60-023-19 1 kg
60-023-20 25 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 60-045-923 Supplier Solstice Supplier No. 312196X1KG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Puriss. p.a., Reag. Ph. Eur.,≥96%

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1305-62-0
Molecular Formula CaH2O2
Molecular Weight (g/mol) 74.09
MDL Number MFCD00010901
InChI Key AXCZMVOFGPJBDE-UHFFFAOYSA-L
Synonym calcium hydroxide, slaked lime, hydrated lime, calcium dihydroxide, calcium ii hydroxide, calcium oxide, hydrated, kalziumhydroxid, loeschkalk, pickling lime, calciumdihydroxid
PubChem CID 6093208
ChEBI CHEBI:31341
IUPAC Name calcium;dihydroxide
SMILES [OH-].[OH-].[Ca++]

Specifications

Quantity 6 x 1 kg
Linear Formula Ca(OH)2
UN Number NONH for all modes of transport
Formula Weight 74.09
Percent Purity ≥96%
Grade puriss. p.a.
Density 2.24g/mL at 25°C
Chemical Name or Material Calcium Hydroxide, Reag. Ph. Eur.
Hazard Statement P261-P280-P305 + P351 + P338
Precautionary Statement H315-H318-H335
EINECSNumber 215-137-3
RTECSNumber EW2800000
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.