Learn More
Calcium Hydroxide TS (Saturated Aqueous Solution), Ricca Chemical
Shop All Ricca Chemical Company Products

Specifications
Specifications
| Color | Colorless |
| CAS Min % | 98.0, 0.29 |
| CAS Max % | 100.0, 0.31 |
| Grade | Laboratory |
| Packaging | Natural Poly Bottle |
| Solvent or Matrix | Saturated Aqueous Solution |
| pH | ∼12.4 |
| Quantity | 4 L |
| Physical Form | Liquid |
| Chemical Name or Material | Calcium Hydroxide, Test Solution |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.