Learn More
Apexbio Technology LLC Calcium Levofolinate 80433-71-2 100mg
SDP
Supplier:  Apexbio Technology LLC B1688100
Encompass_Preferred
Calcium Levofolinate (CAS 80433-71-2) is the calcium salt of levoleucovorin the biologically active enantiomer of folinic acid It functions as a reduced folate cofactor bypassing dihydrofolate reductase inhibition to support DNA and RNA synthesis In biomedical research Calcium Levofolinate is used to modulate the effects of antifolate chemotherapeutic agents such as methotrexate reducing cytotoxicity in normal cells It is also investigated for its ability to enhance the cytotoxic activity of 5-fluorouracil through stabilization of the ternary complex with thymidylate synthase facilitating studies on nucleotide metabolism and chemotherapeutic mechanisms
Catalog No. 50-000-94729
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.