Learn More
CPC Scientific H-Val-Lys-Phe-Gly-Val-Gly-Phe-Lys-OH (trifluoroacetate salt) 5MG
SDP
Supplier:  CPC Scientific CALP002A
Encompass
The peptide acts as a calmodulin agonist, interacts with the calmodulin EF-hand motif, the Ca2+ binding site. CALP-3 activates the phosphodiesterase in the absence of Ca2+ and inhibits cytotoxicity and apoptosis in a Ca2+ mediated manner.ONE-LETTER SEQUENCE: VKFGVGFKMOLECULAR FORMULA: C44H68N10O9MOLECULAR WEIGHT:881.09STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [261969-05-5], RESEARCH AREA: ApoptosisREFERENCES: M.Villain et al., J. Biol. Chem., 275, 2676 (2000); M.K.Manion et al., FASEB J., 14, 1297 (2000); R.Ten Broeke et al., FASEB J., 15, 1831 (2001); R.Ten Broeke et al., Eur. J. Pharmacol., 476, 151 (2003)
Catalog No. 50-210-8866
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.