Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calcium nitrate tetrahydrate, 98%, extra pure

Catalog No. AC447530010
Change view
Click to view available options
Quantity:
1 kg
5 kg
25 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC447530010 1 kg
AC447530050 5 kg
AC447530250 25 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC447530010 Supplier Thermo Scientific Chemicals Supplier No. 447530010
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 13477-34-4 | CaH8N2O10 | 236.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13477-34-4
Molecular Formula CaH8N2O10
Molecular Weight (g/mol) 236.15
InChI Key MWGCGFYACDTFSB-UHFFFAOYSA-N
Synonym calcium nitrate tetrahydrate, calcium nitrate hydrate, puratronic, acmc-1aekc, calcium nitrate tetra hydrate, ca.2no3.4h2o, ca no3 2.4h2o, calcium tetrahydrate dinitronate, calcium nitrate-water 1/4, calcium tetrahydrate dinitrate, calcium 2+ tetrahydrate dinitronate
PubChem CID 16211656
ChEBI CHEBI:86159
IUPAC Name calcium bis(nitric acid) tetrahydrate
SMILES O.[Ca++].[O-][N+]([O-])=O

Specifications

Boiling Point 132.0°C
Melting Point 45.0°C
pH 5.0 to 9.0
Quantity 1 kg
Assay Percent Range 97.5% min. (Complexometry)
Linear Formula Ca(NO3)2·4H2O
Merck Index 15,1684
Solubility Information Solubility in water: 1470g/L (0°C). Other solubilities: soluble in alcohol and acetone
Formula Weight 236.15
Percent Purity 98%
Grade Extra Pure
Specific Gravity 1.86
Density 1.8600g/mL
Chemical Name or Material Calcium nitrate tetrahydrate
Show More Show Less
Hazard Category GHS Signal Word: Danger
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.