Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calcium Nitrate Tetrahydrate, ACS Reagent Grade, Ricca Chemical
SDP

Catalog No. RDCC0100500 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RDCC0100500 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RDCC0100500 Supplier Ricca Chemical Company Supplier No. RDCC0100500B1
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 13477-34-4
Molecular Formula CaH8N2O10
Molecular Weight (g/mol) 236.15
InChI Key MWGCGFYACDTFSB-UHFFFAOYSA-N
IUPAC Name calcium bis(nitric acid) tetrahydrate
SMILES O.[Ca++].[O-][N+]([O-])=O

Specifications

Physical Form Fine Crystals or Crystalline Powder
CAS Max % 103
Quantity 500 g
Packaging White Poly Wide Mouth Bottle
Percent Purity 99 to 103%
Grade ACS
Chemical Name or Material Calcium Nitrate
ShelfLife 84 months
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.