Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calcium nitrate tetrhydrate, Reagent grade, For ACS analysis, ≥99%, MP Biomedicals™

Catalog No. ICN19380183 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
250 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19380183 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN19380183 Supplier MP Biomedicals, Inc Supplier No. 0219380183
Add to Cart
Add to Cart

Chemical Identifiers

CAS 13477-34-4
Molecular Formula CaH8N2O10
Molecular Weight (g/mol) 236.15
InChI Key MWGCGFYACDTFSB-UHFFFAOYSA-N
Synonym calcium nitrate tetrahydrate, calcium nitrate hydrate, puratronic, acmc-1aekc, calcium nitrate tetra hydrate, ca.2no3.4h2o, ca no3 2.4h2o, calcium tetrahydrate dinitronate, calcium nitrate-water 1/4, calcium tetrahydrate dinitrate, calcium 2+ tetrahydrate dinitronate
PubChem CID 16211656
ChEBI CHEBI:86159
IUPAC Name calcium bis(nitric acid) tetrahydrate
SMILES O.[Ca++].[O-][N+]([O-])=O

Specifications

Color White
Physical Form Crystalline Powder
Quantity 250 g
Assay Percent Range ≥99%
Formula Weight 236.2
Grade Reagent
Density 1.86g/mL
Chemical Name or Material Calcium nitrate tetrhydrate, For ACS analysis
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.