Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calcium Phosphate Dibasic, MP Biomedicals™

Catalog No. MP219380490 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 g
500 g
1 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP219380490 500 g
MP219380480 100 g
MP219380491 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. MP219380490 Supplier MP Biomedicals, Inc Supplier No. 0219380490
Add to Cart
Add to Cart

Calcium Phosphate Dibasic, hite Powder

  • It is used in animal feeds, as a mineral supplement, as an antacid, in the manufacturing of glass, in dental products and in fertilizers.
  • It has been used as a dietary supplement, and as an antacid.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7757-93-9
Molecular Formula CaHO4P
Molecular Weight (g/mol) 136.06
MDL Number MFCD00010909
InChI Key FUFJGUQYACFECW-UHFFFAOYSA-L
Synonym Calcium dihydrogenphosphate, Acid calcium phosphate, Calcium biphosphate, Monocalcium orthophosphate
IUPAC Name calcium hydrogen phosphate
SMILES [Ca++].OP([O-])([O-])=O

Specifications

Melting Point ∼370°C with decomposition (literature)
Color White
Density ca. 2.3g/cm3 at 20°C (Lit.)
Boiling Point Does not boil (Literature)
Quantity 500 g
Solubility Information Soluble in 1 M Hydrochloric acid (100mg/mL - slightly hazy, colorless to slight yellowish-green solution).
Physical Form Powder
Assay 0.9829
storageNote1 Store at Room Temperature (15-30°C).
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.