Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calcium Phthalate Hydrate 98.0+%, TCI America™
SDP

Catalog No. C200325G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C200325G 25 g
C20035G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C200325G Supplier TCI America Supplier No. C200325G

Chemical Identifiers

CAS 5793-85-1
Molecular Formula C8H4CaO4
Molecular Weight (g/mol) 204.19
MDL Number MFCD08276328
InChI Key PYSZASIZWHHPHJ-UHFFFAOYSA-L
Synonym Phthalic Acid Calcium Salt
PubChem CID 91659099
IUPAC Name calcium benzene-1,2-dicarboxylate
SMILES [Ca++].[O-]C(=O)C1=CC=CC=C1C([O-])=O

Specifications

Color White
Quantity 25 g
Formula Weight 204.19
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Calcium Phthalate Hydrate
EINECSNumber (3)-1272
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.