Learn More
Sigma Organic Chemistry calcium salicylate | 500mg | MFCD00045807
Supplier: Sigma Organic Chemistry CDS003348500MG

calcium salicylate , 500mg
About this Item:
MDL #: MFCD00045807
Molecular Weight: 314.3
UNSPSC Code: 12352200
Chemical Formula: C14H10CaO6
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.