Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Calcium Sensing Receptor Monoclonal Antibody (HL 1499)
GREENER_CHOICE

Catalog No. 01950209
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-950-209 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-950-209 Supplier Invitrogen™ Supplier No. MA116940
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Suggested positive control: antigen standard for CASR (transient overexpression lysate), human kidney protein.

Senses changes in the extracellular concentration of calcium ions. The activity of this receptor is mediated by a G- protein that activates a phosphatidylinositol-calcium second messenger system.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Calcium Sensing Receptor
Applications ELISA, Flow Cytometry, Immunoprecipitation, Radioimmune Assays (RIA), Western Blot
Classification Monoclonal
Clone HL 1499
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene CASR
Gene Accession No. P41180, P48442, Q9QY96
Gene Alias BoPCaR1; Ca sensing receptor; Ca(2+)-sensing receptor; calcium sensing receptor; calcium-sensing receptor; Calcium-sensing receptor (hypocalciuric hypercalcemia 1 severe neonatal hyperparathyroidism); Calcium-sensing receptor (hypocalciuric hypercalcemia 1, severe neonatal hyperparathyroidism); CaR; CaSR; CaSR antibody; cation sensing receptor; EIG8; Extracellular calcium-sensing receptor; FHH; FIH; G protein coupled receptor, family C, group 2, member A; G protein-coupled receptor, family C, group 2, member A; Gprc2a; HHC; HHC1; HYPOC1; LOW QUALITY PROTEIN: extracellular calcium-sensing receptor; NSHPT; parathyroid Ca(2+)-sensing receptor 1; parathyroid cell calcium-sensing receptor; Parathyroid cell calcium-sensing receptor 1; PCaR1
Gene Symbols CASR
Host Species Mouse
Immunogen N-terminal region of human Calcium Sensing Receptor.
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100341543, 12374, 24247, 846
Target Species Human, Mouse, Rabbit, Rat
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.