Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calcium Silicate, NF, Spectrum™ Chemical
SDP

Catalog No. 18601379 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
1 kg
8 kg
15 kg
Packaging:
Fiber Drum
Poly Pail
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-601-379 8 kg Fiber Drum
18-601-380 15 kg Fiber Drum
18-601-376 1 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18-601-379 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. C16438KG
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart

C1643, 1344-95-2, CaO·SiO2

Spectrum™ Chemical Calcium Silicate, NF , also known as calcium orthosilicate, is used as an anticaking agent in pharmaceutical preparations. All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 1344-95-2
Molecular Formula CaO3Si
Molecular Weight (g/mol) 116.16
InChI Key OYACROKNLOSFPA-UHFFFAOYSA-N
IUPAC Name calcium oxosilanebis(olate)
SMILES [Ca++].[O-][Si]([O-])=O

Specifications

CAS Max % 100%
pH 8.4-11.2
Quantity 8 kg
Grade NF
Loss on Ignition 20%
Packaging Fiber Drum
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.