Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calcium silicide, tech.

Catalog No. p-7049552
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1467636 500 g
AA1467622 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA1467636 Supplier Thermo Scientific Chemicals Supplier No. 01467636
Only null left
Add to Cart

CAS: 12013-56-8 | CaSi2 | 96.248 g/mol

It is an important raw material used in Pharmaceutical intermediates. It is an ideal compound of deoxidizer and desulfurizer which can be used in production of many types of stainless steel. It is also used as inoculants and additive in cast iron.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12013-56-8
Molecular Formula CaSi2
Molecular Weight (g/mol) 96.248
MDL Number MFCD00015977
InChI Key XXCKHUMTXOCLPT-UHFFFAOYSA-N
Synonym calciumdisilicid, ? 1-silylcalcio-? 1-silanylidyne
PubChem CID 13848147
IUPAC Name calcium;silicon(1-)
SMILES [Si-].[Si-].[Ca+2]

Specifications

Melting Point ∼1000°C
Color Gray
Physical Form Powder
Quantity 500 g
UN Number UN1405
Solubility Information Insoluble in cold water, decomposes in hot water
Formula Weight 96.25
Grade Technical
Concentration or Composition (by Analyte or Components) Ca ≈30%, may contain up to 5% Fe
Sensitivity Moisture sensitive
Density 2.5 g/mL
Chemical Name or Material Calcium silicide
Show More Show Less
Hazard Category H228
Hazard Statement GHS H Statement
H261
In contact with water releases flammable gas.
Precautionary Statement P210-P240-P241-P280-P370+P378q
DOTInformation Transport Hazard Class: 4.3; Packing Group: II; Proper Shipping Name: CALCIUM SILICIDE
EINECSNumber 234-588-7
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.