Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calcium Sulfate Hemihydrate, Purified (Plaster of Paris), Reagents

Catalog No. 01337327
Change view
Click to view available options
Quantity:
500 g
2 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-337-327 500 g
01-337-328 2 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01-337-327 Supplier Reagents Holdings Llc Supplier No. C2168000500A5
Add to Cart
Add to Cart

Calcium Sulfate Hemihydrate, Purified (Plaster of Paris), Technical, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 10034-76-1
Molecular Formula Ca2H2O9S2
Molecular Weight (g/mol) 290.28
InChI Key ZOMBKNNSYQHRCA-UHFFFAOYSA-J
Synonym Plaster of Paris, Gypsum hemihydrate
IUPAC Name dicalcium hydrate disulfate
SMILES O.[Ca++].[Ca++].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O

Specifications

Color Colorless/White
Physical Form Solid
Quantity 500 g
Grade Technical
Packaging Poly Bottle
Specific Gravity 2.3
Chemical Name or Material Calcium Sulfate Hemihydrate, Purified (Plaster of Paris)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.