Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calcium Sulfate Solution R, Saturated, Ricca Chemical
SDP

Catalog No. 182316 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
120 mL
500 mL
Packaging:
Natural Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
1823-16 500 mL Natural Poly Bottle
R1823-4 120 mL Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 1823-16 Supplier Ricca Chemical Company Supplier No. 182316
Add to Cart
Add to Cart

Chemical Identifiers

CAS 10101-41-4 , 7732-18-5
Molecular Formula CaH4O6S
Molecular Weight (g/mol) 172.16
MDL Number MFCD00149625
InChI Key PASHVRUKOFIRIK-UHFFFAOYSA-L
Synonym calcium sulfate dihydrate, phosphogypsum, landplaster, annaline, compactrol, gips, gypsite, primoplast, satinite, gypsum stone
PubChem CID 24928
ChEBI CHEBI:32583
IUPAC Name calcium dihydrate sulfate
SMILES O.O.[Ca++].[O-]S([O-])(=O)=O

Specifications

Color Colorless
Grade EP
Packaging Natural Poly Bottle
Solvent or Matrix Saturated Aqueous Solution
Quantity 500 mL
Physical Form Liquid
Chemical Name or Material Calcium Sulfate Solution, Reagent
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.