Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calcium titanium oxide, 99+% (metals basis)

Catalog No. AA11397A3
Change view
Click to view available options
Quantity:
500 g
2 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA11397A3 2 kg
AA1139736 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA11397A3 Supplier Thermo Scientific Chemicals Supplier No. 011397A3
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 12049-50-2 | CaO3Ti | 135.94 g/mol

Calcium titanium oxide is used to prepare the ferrotitanium alloys and its structure is used to study the other compounds such as barium titanate.It is used to make the pure substances at low temperature.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12049-50-2
Molecular Formula CaO3Ti
Molecular Weight (g/mol) 135.94
MDL Number MFCD00015981
InChI Key WEUCVIBPSSMHJG-UHFFFAOYSA-N
Synonym calcium titanate, titaniumcalciumoxide, calciumtitaniumtrioxide, ca.o3ti, titanatecalcium 1:1, calcium dioxido oxo titanium, calcium oxotitaniumbis olate, calcium 2+ oxotitaniumbis olate, titanate tio32-,calcium 1:1, calcium titanate, nanopowder, <100 nm particle size bet trace metals basis
PubChem CID 16212381
IUPAC Name calcium;dioxido(oxo)titanium
SMILES [O--].[O--].[O--].[Ca++].[Ti+4]

Specifications

Melting Point 1975°C
Color Beige
Physical Form Powder
Quantity 2 kg
Odor Odorless
Assay Percent Range >99% (Metals basis)
Density 4.10 g/cm3
Solubility Information Insoluble in water. Soluble in oirganic and inorganic solution.
Formula Weight 135.98
Chemical Name or Material Calcium titanium oxide
EINECSNumber 234-988-1
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.