Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calcium, Turnings 99.5%, Thermo Scientific™

Catalog No. AC201385000
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC201385000 500 g
AC201381000 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC201385000 Supplier Thermo Scientific Chemicals Supplier No. 201385000

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-70-2
Molecular Formula Ca
Molecular Weight (g/mol) 40.08
MDL Number MFCD00010897 MFCD00085314
InChI Key OYPRJOBELJOOCE-UHFFFAOYSA-N
Synonym unii-sy7q814vup, hsdb 273, sy7q814vup, compounds, elemental, calcium, elemental, kalzium, atom, ingot, chelate
PubChem CID 5460341
ChEBI CHEBI:29320
IUPAC Name calcium
SMILES [Ca]

Specifications

Density 1.54
Quantity 500 g
Form Turnings
Linear Formula Ca
Fieser 05,89; 09,1634; 14,71
Merck Index 15, 1648
Formula Weight 40.07
Percent Purity ≥99.4% (Ca)
Chemical Name or Material Calcium, 99.5%
Hazard Category GHS Signal Word: Danger
EINECSNumber 231-179-5
TSCA TSCA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.