Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CALCRL Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA597108 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA597108 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA597108 Supplier Invitrogen™ Supplier No. PA597108
  • $575.00
    $472.65 / Each of 1

    Save $102.35

    18% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit Polyclonal Antibody

Positive Samples: A-549, MCF7, Mouse kidney, Mouse liver, Rat liver; Cellular Location: Cell membrane, Multi-pass membrane protein.

CRLR is a receptor for calcitonin gene-related peptide type 1. The activity of CRLR is mediated by G proteins which activate adenylyl cyclase. CRLR expression has been reported in brain, lung, blood vessel, liver, and intestinal tract. ESTs have been isolated from B-cell/lung/testis, bome marrow, embryo, lung and synovium libraries.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CALCRL
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.52 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene CALCRL
Gene Accession No. Q16602, Q63118, Q9R1W5
Gene Alias AV071593; Calcitonin gene-related peptide type 1 receptor; calcitonin receptor like receptor; calcitonin receptor-like; calcitonin receptor-like receptor; CALCRL; CGRP receptor; CGRP type 1 receptor; Cgrp1 receptor; CGRPR; CLR; CRLR; RATCRLR; RNCLR
Gene Symbols CALCRL
Host Species Rabbit
Immunogen Synthetic peptide.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10203, 25029, 54598
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.