Learn More
Description
MA5-11626 targets Caldesmon HMW in IHC (P), IF and WB applications and shows reactivity with human and mouse samples. The MA5-11626 immunogen is crude human uterus extract.
Specifications
Specifications
| Antigen | Caldesmon HMW |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | h-CALD |
| Concentration | 0.2 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.05% BSA and 0.05% sodium azide; pH 7.4 |
| Gene | Cald1 |
| Gene Accession No. | Q05682, Q62736 |
| Gene Alias | 4833423D12Rik; AI195384; AV071549; AW536160; C920027I18Rik; CAD; Cald1; Caldesmon; caldesmon 1; CDM; HCAD; H-CAD; h-caldesmon; LCAD; L-CAD; L-caldesmon; MGC21352; NAG22; Non-muscle caldesmon; OTTHUMP00000207950; OTTHUMP00000207953; OTTHUMP00000207954; OTTHUMP00000207955; OTTHUMP00000214473; smooth muscle caldesmon; testis secretory sperm-binding protein Li 227n |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
