Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Honeywell Analytics™ CalGas Calibration Gases

Catalog No. 19806876
Encompass
Encompass_Preferred
Change view
Click to view available options
Components Measured:
Propane (LEL), Carbon Monoxide, Hydrogen Sulfide
Capacity (Metric):
34 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Components Measured Capacity (Metric)
19-806-876 Propane (LEL), Carbon Monoxide, Hydrogen Sulfide 34 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-806-876 Supplier Honeywell Analytics™ Supplier No. 549049E
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Choose from both multigas mixtures and single-gas calibration gases

  • Calibrates gas detectors to ensure accuracy
  • Multigas mixtures presented are not for use with CO+ or Tox1 sensors

Order Info

  • Among single-gas calibration gases, Cat. Nos. 19-806-863 and 19-806-880 are for Mighty-Tox series bump test only; 19-806-865 is for use with Biosystems 12-022 regulator (19-806-687) only.
  • For information about additional gases not offered here, contact your Fisher Safety Customer Service Representative at 1-800-772-6733.

TRUSTED_SUSTAINABILITY

Specifications

Capacity (English) 8.98 gal.
Capacity (Metric) 34 L
Concentration Propane (LEL 50%), carbon monoxide 50ppm, hydrogen sulfide 25ppm
Components Measured Propane (LEL), Carbon Monoxide, Hydrogen Sulfide
Type Calibration Gas
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.