Learn More
Thermco Products Calibrated Smart Probe
Catalog No. 04500051
Change view
Click to view available options
For Use With (Equipment):
Data Loggers
Catalog No. For Use With (Equipment)
04-500-051 Data Loggers
1 options
Catalog No. 04-500-051 Supplier Thermco Products Supplier No. LTCP30
Add to Cart
Add to Cart
These precise reliable probes compatible with UTRED30-16 and UTRED30-WiFi Dual-Channel loggers measure temperatures from -40°C to +99°C (-40°F to 210°F).
  • Designed for seamless integration, allowing probes to be swapped and calibrated independently to minimize logging downtime and eliminate the need to return or relocate the logger for calibration.
  • Plug-and-play, hot-swappable design enables easy probe replacement while expired probes can be sent for re-calibration, ensuring uninterrupted logging.
  • Ensures continuous and accurate temperature monitoring for vaccine storage and other critical applications, supporting compliance and operational efficiency.
  • Precise probes with a temperature range from -40°C to +99°C (-40°F to +210°F).
  • Hot-swappable design for minimal downtime.
  • Independent calibration with no logger relocation required.
  • Compatible with UTRED30-16 and UTRED30-WiFi loggers.
  • Ensures continuous and reliable temperature monitoring.
Specifications
Type Probe
For Use With (Equipment) Data Loggers