Description
- Calibration certificate
- For temperature
Specifications
Specifications
| For Use With (Equipment) | BINDER Chambers Series KB ECO |
| Description | Calibration certificate, temperature - temperature measurement incl. certificate, 9 measuring points (at specified temperature) |
| Duration | 60 to 120 min. |
| Calibration Type | 9 P Temperature |
| Product Type | Calibration Certificate |