Learn More
BINDER™ Calibration Certificate for BINDER™ KB ECO Series
SDP
Catalog No. 08112080 Shop All BINDER Inc. Products
Change view
Click to view available options
Calibration Type:
1 P Expanded Temperature
1 P Temperature
18 P Temperature
27 P Temperature
9 P Temperature
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Calibration Type
08-112-080 18 P Temperature
08-112-077 1 P Temperature
08-112-078 1 P Expanded Temperature
08-112-079 9 P Temperature
08-112-081 27 P Temperature
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 08-112-080 Supplier BINDER™ Supplier No. 80122109
Add to Cart
Add to Cart
The Calibration Certificate for BINDER KB ECO Series is an option for KB ECO Series Refrigerated Incubators with environmentally friendly Thermoelectric Cooling.
  • Calibration certificate
  • For temperature
Specifications
For Use With (Equipment) BINDER Chambers Series KB ECO
Description Calibration certificate, temperature - temperature measurement incl. certificate, 15- 18 measuring points (at specified temperature)
Duration 60 to 120 min.
Calibration Type 18 P Temperature
Product Type Calibration Certificate