Learn More
Sigma Aldrich Fine Chemicals Biosciences AldrichR desiccator cabine1EA
Supplier: Sigma Aldrich Fine Chemicals Biosciences Z2681351EA
Aldrich desiccator cabinets are dust and moisture-free storage for refrigerators freezers and at room temperature. The clear walls top and base are 1/4 in. thick in acrylic for 360 visability.Gasketed air-tight doors are 3/8Ain. thick acrylic with full length hinges and SS twist locking latches. Clear acrylic deep slide-in trays with perforated bottoms for proper circulation within the cabinet. Dark bronze acrylic provides maximum protection for light-sensitive materials.