Learn More
LABSYSTEMS DIAGNOSTICS OY Calibrators for Vitamin A/E assay
Supplier:  LABSYSTEMS DIAGNOSTICS OY 3469877
Encompass_Preferred
VAE-calibration standard is intended to be used in clinical laboratories for the quantitative determination of vitamin A and E in serum and plasma to detect deficiencies or hypervitaminosis. The calibrators are prepared using human serum and in a lyophilized form
Catalog No. NC2865512