Learn More
DWK Life Sciences Calibrex Flow Control Stopcock for Bottle Top Dispensers
SDP
Catalog No. 13710933 Shop All DWK Life Sciences Products
Change view
Click to view available options
For Use With (Equipment):
Calibrex 525/530 Bottle-top Dispensers (25, 50 and 100mL)
Catalog No. For Use With (Equipment)
13-710-933 Calibrex 525/530 Bottle-top Dispensers (25, 50 and 100mL)
1 options
Catalog No. 13-710-933 Supplier DWK Life Sciences Supplier No. W844114
Add to Cart
Add to Cart
Interchangeable flow control stopcock for all Calibrex™ 525 and 530 bottle top dispensers.
A flow control stopcock enables liquid priming and recycling without reagent loss or contamination.
TRUSTED_SUSTAINABILITY
Specifications
Product Type Flow Control Stopcock
For Use With (Equipment) Calibrex 525/530 Bottle-top Dispensers (25, 50 and 100mL)