Learn More
DWK LIFE SCIENCES™ Calibrex™ solutae 530 Bottle Top Dispenser with Flow Control Stopcock
Shop All DWK Life Sciences Products

Description
- PFA-coated plunger prevents plunger and barrel from seizing, enabling trouble-free distribution of salt solutions, weak and strong acids, and bases
- Flow control stopcock enables liquid priming and recycling without reagent loss or contamination
- Permanent fluid path visibility
- Integrated calibration system
- Choice of adjustment button included with each unit
- Long-lasting performance stability
- Autoclavable at 121°C / 250°F fully assembled
- Two-year warranty
Specifications
Specifications
| Product Type | Bottletop Dispenser |
| Autoclavable | Autoclavable |
| Channels | 1 |
| Material | Borosilicate Glass |
| Volume (Metric) | 1 to 10 mL |
| For Use With (Equipment) | Fits most laboratory bottles directly or with supplied adapters |
| Includes | Flow Control Stopcock, PFA-coated plunger and integrated calibration |
| Increments | 0.2 mL |
| Model | Calibrex solutae 530 |
| Warranty | 2 Years |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.