Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Calicivirus Monoclonal Antibody (FCV1-43)
GREENER_CHOICE

Catalog No. PIMA182322 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
250 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA182322 250 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA182322 Supplier Invitrogen™ Supplier No. MA182322
  • $861.00
    $699.65 / Each of 1

    Save $161.35

    19% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Mouse Monoclonal Antibody

Heat-mediated antigen retrieval is recommended for the staining of paraffin sections. Mouse anti Feline calicivirus antibody, clone FCV1-43 recognizes feline calicivirus, and does not cross-react with FIV or FeLV.

TRUSTED_SUSTAINABILITY

Specifications

Antigen Calicivirus
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone FCV1-43
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with <0.1% sodium azide; pH 7.2
Gene Alias Feline calicivirus
Host Species Mouse
Immunogen Proprietary Immunogen.
Purification Method Protein A
Quantity 250 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.