Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Calmodulin 2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5121166
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5121166 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5121166 Supplier Invitrogen™ Supplier No. PA5121166
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive test controls include: NIH/3T3, MCF7, BT-474, NCI-H460, Mouse brain. The target is usually found in the following locations: Cytoplasm, cytoskeleton, spindle, spindle pole. Immunogen sequence: MADQLTEEQI AEFKEAFSLF DKDGDGTITT KELGTVMRSL GQNPTEAELQ DMINEVDADG NGTIDFPEFL TMMARKMKDT DSEEEIREAF RVFDKDGNGY ISAAELRHVM TNLGEKLTDE EVDEMIREAD IDGDGQVNYE EFVQMMTAK.

Calmodulin 2 (CALM2) is one of the three human genes encoding calmodulin, a highly conserved calcium-binding messenger protein critical for mediating various cellular processes. Calmodulin proteins are composed of two pairs of EF-hand motifs located at both the N- and C-termini, connected by a flexible central linker. Each EF-hand motif binds calcium ions, with the C-terminus possessing two high-affinity Ca2+ binding sites, while the N-terminus contains two low-affinity sites. Upon binding calcium, calmodulin undergoes a conformational change that enables it to interact with and regulate numerous target proteins, including kinases, phosphatases, ion channels, and other enzymes. Specifically, calcium/calmodulin (CaM) signaling pathways are involved in critical cellular functions such as muscle contraction, neurotransmitter release, and cellular growth. The interaction of CALM2 with different targets is essential for translating transient changes in intracellular calcium levels into specific cellular responses, making it a pivotal player in calcium signaling.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Calmodulin 2
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.08 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene CALM2
Gene Accession No. P0DP24, P0DP27
Gene Alias 1500001E21Rik; AL024017; Calm; Calm1; CALM2; Calm3; CALML2; calmodulin; calmodulin 2; calmodulin 2 (phosphorylase kinase, delta); Calmodulin-1; Calmodulin-2; Calmodulin-3; CAM; CAM1; Cam2; CAM3; CAMB; Camc; CaMI; CaMII; CaMIII; LP7057 protein; LQT15; PHKD; PHKD2; phosphorylase kinase delta; phosphorylase kinase subunit delta; prepro-calmodulin 2
Gene Symbols CALM2
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-149 of human CALM2 (NP_001734.1).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12314, 805
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.