Learn More
CPC Scientific H-Leu-Lys-Lys-Phe-Asn-Ala-Arg-Arg-Lys-Leu-Lys-Gly-Ala-Ile-Leu-Thr-Thr-Met-Leu-Ala-OH (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific PKCP008A
Encompass
Potent calmodulin antagonist that is a Ca2+/Calmodulin-dependent protein kinase II inhibitor.ONE-LETTER SEQUENCE: LKKFNARRKLKGAILTTMLAMOLECULAR FORMULA: C103H185N31O24S1MOLECULAR WEIGHT:2273.83STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [115044-69-4], RESEARCH AREA: Protein Kinases and PhosphatasesREFERENCES: M.E. Payne et al., JBC, 263, 7190 (1989)
Catalog No. 50-211-0161
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.