Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Calnexin Monoclonal Antibody (AF18), Alexa Fluor™ 488
GREENER_CHOICE

Catalog No. PIMA3027A48 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA3027A48 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA3027A48 Supplier Invitrogen™ Supplier No. MA3027A488
  • $548.00
    $440.65 / Each of 1

    Save $107.35

    20% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Mouse Monoclonal Antibody

MA3027A488 detects calnexin from human and mouse tissues. The MA3027A488 antigen is Focus human hepatoma cell lysate.

Calnexin, also referred to as IP90, p88 and p90, is an approximately 90 kDa integral membrane protein of the endoplasmic reticulum (ER). Many resident ER proteins act as molecular chaperones and participate in the proper folding of polypeptides and their assembly into multisubunit proteins. Studies indicate that calnexin associates with the major histocompatibility complex (MHC) class I heavy chains, partial complexes of the T cell receptor and B cell membrane immunoglobulin, but not with completed receptor complexes. It has been shown that calnexin is a chaperone that retains incompletely or improperly folded proteins in the ER. The sequence Lys-Asp-Glu-Leu (KDEL) or a closely related sequence, is present at the carboxy-terminus of soluble ER resident proteins such as GRP78 and GRP94 and protein disulfide isomerase. Integral membrane ER resident proteins, like calnexin, often lack this KDEL sequence but contain positively charged cytosolic residues that ensure ER retention. Calnexin contains a large ER luminal domain (461 amino acids), a transmembrane segment (22 amino acids), and a cytoplasmic tail (89 amino acids).
TRUSTED_SUSTAINABILITY

Specifications

Antigen Calnexin
Applications Immunocytochemistry
Classification Monoclonal
Clone AF18
Concentration 1 mg/mL
Conjugate Alexa Fluor 488
Formulation PBS with 1mg/mL BSA and 0.05% sodium azide; pH 7.4
Gene CANX
Gene Accession No. P27824, P35564
Gene Alias 1110069N15Rik; AI988026; At5g07340; At5g61790; calmegin; calnexin; canx; canx-a; canx-b; clgn; clnx; cnx; CNX1; CNX2; D11Ertd153e; FLJ26570; hypothetical protein; IP90; MAC9.11; Major histocompatibility complex class I antigen-binding protein p88; p88; P90; PP90; pp90 precursor
Gene Symbols CANX
Host Species Mouse
Immunogen Focus human hepatoma cell lysate.
Purification Method Protein A
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12330, 821
Target Species Human, Mouse
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.