Learn More
Description
Keep as concentrated solution. Predicted reactivity: Mouse (93%), Rat (93%), Zebrafish (82%), Xenopus laevis (89%), Dog (94%), Pig (94%), Chicken (91%), Bovine (94%). Positive Control: mouse liver, PC-12, Rat2, 293T, A431, HeLa, HepG2. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Specifications
Specifications
| Antigen | Calnexin |
| Applications | Dot Blot, Immunoprecipitation, Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | GT1563 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative |
| Gene | CANX |
| Gene Accession No. | P27824, P35564, P35565 |
| Gene Alias | 1110069N15Rik; AI988026; calmegin; calnexin; canx; canx-a; canx-b; clgn; clnx; cnx; D11Ertd153e; FLJ26570; hypothetical protein; IP90; Major histocompatibility complex class I antigen-binding protein p88; p88; P90; PP90; pp90 precursor |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
