Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Calpain 1 Native Protein
SDP

Catalog No. NB002065 Shop All Novus Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB002065 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB002065 Supplier Novus Biologicals™ Supplier No. NBP199920100UG

missing translation for 'validMainframeMsg'

Highly purified. Generating reliable and reproducible results. Applications: SDS-Page, Bioactivity

Native Calpain 1

Source:Human Erythrocyte

Amino Acid Sequence:(P07384)

Specifications

For Use With (Application) Bioactivity, SDS-PAGE
Formulation 20 mM imidazole-HCl (pH 6.8), 5 mM B-mercaptoethanol, 1 mM EDTA, 1 mM EGTA, 30% glycerol
Gene ID (Entrez) 823
Molecular Weight (g/mol) 112 kDa
Quantity 100 μg
Source Human Erythrocytes
Storage Requirements Store at -20 to -80C. Avoid freeze-thaw cycles.
Gene Alias Calcium-activated neutral proteinase 1, calpain 1, (mu/I) large subunit, Calpain mu-type, calpain, large polypeptide L1, calpain-1 catalytic subunit, Calpain-1 large subunit, CANP, CANP 1, CANPL1EC 3.4.22.52, Cell proliferation-inducing gene 30 protein, cell proliferation-inducing protein 30, EC 3.4.22, Micromolar-calpain, muCANPCANP1, muCL
Gene Symbol CAPN1
Buffer 20 mM imidazole-HCl (pH 6.8), 5 mM B-mercaptoethanol, 1 mM EDTA, 1 mM EGTA, 30% glycerol
Purity or Quality Grade >95%, by SDS-PAGE
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.