Learn More
Invitrogen™ Calpain 11 Polyclonal Antibody

Description
Antibody detects endogenous levels of total CAPN11.
Specifications
Specifications
| Antigen | Calpain 11 |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 |
| Gene | CAPN11 |
| Gene Accession No. | Q9UMQ6 |
| Gene Alias | calcium-activated neutral proteinase 11; calcium-activated neutral proteinase 11 {ECO:0000250; calcium-dependent thiol protease; calpain 11; calpain 11 {ECO:0000312; calpain11; Calpain-11; calpain-11; LOW QUALITY PROTEIN: calpain-11; CANP 11; CANP 11 {ECO:0000250; Capn11; capn11 {ECO:0000312; EMBL:AAH97256.1}; HIP2; LIG; RGD:1302946}; UniProtKB:Q9UMQ6} |
| Gene Symbols | CAPN11 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.