Learn More
Invitrogen™ Calpain 2 Recombinant Rabbit Monoclonal Antibody (9Y7I1)

Description
Immunogen sequence: LWTKIQKYQK IYREIDVDRS GTMNSYEMRK ALEEAGFKMP CQLHQVIVAR FADDQLIIDF DNFVRCLVRL ETLFKIFKQL DPENTGTIEL DLISWLCFSV L.
Specifications
Specifications
| Antigen | Calpain 2 |
| Applications | ELISA, Western Blot |
| Classification | Recombinant Monoclonal |
| Clone | 9Y7I1 |
| Concentration | 1.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | CAPN2 |
| Gene Accession No. | O08529, P17655, Q07009 |
| Gene Alias | 80 kDa M-calpain subunit; AI326419; Calcium-activated neutral proteinase 2; calcium-dependent protease; CALP80; calpain 2; calpain 2, (m/II) large subunit; calpain 2, large [catalytic] subunit; calpain 2, large subunit; calpain II; calpain large polypeptide L2; Calpain M-type; calpain, large polypeptide L2; calpain-2 catalytic subunit; Calpain-2 large subunit; CANP 2; CANP2; CANPL2; CANPml; Capa2; Capa-2; CAPN2; cysteine proteinase; m-calpain; m-calpain 80 kDa large subunit; m-calpain large subunit; m-calpin; mCANP; millimolar-calpain |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.