Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Calpain Inhibitor II, 95+%

Catalog No. AAJ62491MB
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ62491MB 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ62491MB Supplier Thermo Scientific Chemicals Supplier No. J62491MB
Get 15% Off Thermo Scientific Reagents
Use promo code REAGENTS15
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

An inhibitor of calpains I and II

ALLM (Calpain Inhibitor) is a cell-permeable, peptide aldehyde inhibitor of calpain I and calpain II. ALLM acts as a very potent inhibitor of cathepsin L and the strongest inhibitor of cathepsin B amongst the peptide aldehydes. This compound inhibits the processing of malaria aspartic hemoglobinases plasmepsins I and II in vitro. ALLM also inhibits other neutral cysteine proteases, and activation-induced programmed cell death. It restores defective immune responses in HIV+ donors. ALLM blocks nitric oxide production by activated macrophages by interfering with transcription of the inducible nitric oxide synthase gene. ALLM also acts as a weak inhibitor of proteasome.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 136632-32-1
Molecular Formula C19H35N3O4S
Synonym N-Ac-Leu-Leu-Methioninal; ALLM

Specifications

Quantity 25 mg
Description White
Content And Storage -30°C to -10°C
For Use With (Application) An inhibitor of calpains I and II
Formula Weight 401.60g/mol
Solubility Soluble in ethanol at 20mg/ml
Product Type Signal Transduction Reagent
Form Powder
Assay ≥95%

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.