Learn More
Invitrogen™ Calpain S1/S2 Monoclonal Antibody (28F3)

Description
This antibody specifically recognizes the 28 to 30 kD small subunit of Calpain 1 and Calpain 2 (mu calpain and m calpain). It recognizes both native and denatured protein in ELISA, Western Blot and Immunoprecipitation.
Specifications
Specifications
| Antigen | Calpain S1/S2 |
| Applications | ELISA, Immunoprecipitation, Western Blot |
| Classification | Monoclonal |
| Clone | 28F3 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 3mM sodium azide; pH 7.5 |
| Gene | CAPNS1 |
| Gene Accession No. | P04632, P13135, Q96L46 |
| Gene Alias | 2310005G05Rik; 30K-2; calcium-activated neutral proteinase 2; Calcium-activated neutral proteinase small subunit; Calcium-dependent protease small subunit; Calcium-dependent protease small subunit 1; calcium-dependent protease small subunit 2; calcium-dependent protease, small subunit; calpain 2; calpain 2, (m/II) large subunit; calpain 2, large [catalytic] subunit; calpain 2, large subunit; calpain 4; calpain 4, small subunit (30K); calpain II regulatory subunit; calpain II regulatory subunit (EC 3.4.22.17); calpain large polypeptide L2; Calpain M-type; calpain regulatory subunit; Calpain small subunit 1; calpain small subunit 2; calpain, large polypeptide L2; calpain, small polypeptide; calpain, small subunit 1; calpain, small subunit 2; calpain-2 catalytic subunit; Calpain-2 large subunit; CALPAIN4; CANP; CANP 2; CANP small subunit; CANP2; CANPL2; CANPml; CANPS; Capa4; Capa-4; CAPN2; Capn4; CAPNS; CAPNS1; Capns2; CDPS; CSS1; CSS2; D7Ertd146e; M-calpain; mCANP; Millimolar-calpain |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.