Learn More
Apexbio Technology LLC Calpeptin 117591-20-5 50mg
SDP
Supplier:  Apexbio Technology LLC A441150
Encompass_Preferred
Calpeptin (CAS 117591-20-5) is a potent inhibitor of calpains calcium-dependent intracellular cysteine proteases involved in cellular differentiation proliferation and apoptosis It inhibits human calpain 1 with an IC50 of approximately 5 nM In vitro studies demonstrate that Calpeptin suppresses the production of TGF- 1 IL-6 and angiopoietin-1 as well as collagen synthesis in lung fibroblasts In mouse models of bleomycin-induced pulmonary fibrosis Calpeptin treatment reduces pulmonary tissue fibrosis accompanied by decreased expression of IL-6 TGF- 1 angiopoietin-1 and collagen type Ia1 mRNA Calpeptin serves as a valuable tool in fibrosis-related research
Catalog No. 50-000-61854
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.