Learn More
Description
This antibody raised against the specific region of human CNN1, thus it does not cross-react with CNN2 or CNN3.
Specifications
Specifications
| Antigen | Calponin |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 0.33 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 |
| Gene | CNN1 |
| Gene Accession No. | P51911, Q08091 |
| Gene Alias | Basic calponin, Calponin, Calponin 1, Calponin H1, smooth muscle, CNN1, Sm Calp, SMCC |
| Gene Symbols | CNN1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
