Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GENOVA DIAGNOSTICS CALP0170RUO, QEV2 / 70NM LEGACY, ELISA kit, 2 PACK, Calprotectin assay, 5-500 ng/mL range, 96 well pre-coated plate, research use only

Supplier:  GENOVA DIAGNOSTICS CALP0170RUO

Encompass

The CALP0170RUO is an ELISA-based kit designed for the quantitative detection of Calprotectin, an important biomarker for inflammation and infections. This kit supports research across human and non-human primate models, providing flexibility in sample types and processing methods. It accommodates both manual and automated workflows, ensuring efficiency in laboratory settings.

  • Assay target is Calprotectin.
  • Quantitative read-out allows precise measurement.
  • Detection range from 5 to 500 ng/mL.
  • Assay can be completed in approximately 2 hours.
  • Detection system uses ALP/pNPP read at 405 nm.
  • Regulatory status is Research Use Only.
  • Validated sample types include EDTA plasma and faeces.
  • Species reactivity for human and non-human primates.
  • Utilizes a 96 well pre-coated ELISA plate format.
  • Supports up to 80 wells for sample processing.
  • Compatible with Dynex DS2 automated systems.

Catalog No. NC2101721


May include imposed supplier surcharges.
Only null left
Add to Cart
This item is not returnable. View return policy

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.