Learn More
Calreticulin Antibody (CR213-2AG), FITC, R&D Systems™

Description
Specifications
Specifications
| Antigen | Calreticulin |
| Applications | ELISA, Western Blot |
| Classification | Monoclonal |
| Clone | CR213-2AG |
| Conjugate | FITC |
| Dilution | ELISA |
| Formulation | PBS |
| Gene Alias | autoantigen Ro, CALR, calregulin, calreticulin, cC1qR, CRP55, CRT, Endoplasmic reticulum resident protein 60, epididymis secretory sperm binding protein Li 99n, ERp60, FLJ26680, grp60, HACBP, HEL-S-99n, RO, Sicca syndrome antigen A (autoantigen Ro; calreticulin), SSA |
| Host Species | Mouse |
| Immunogen | Recombinant human calreticulin. |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.