Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Calreticulin Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA1902A Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA1902A 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA1902A Supplier Invitrogen™ Supplier No. PA1902A
  • $720.00
    $500.65 / Each of 1

    Save $219.35

    30% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Chicken Polyclonal Antibody

PA1-902A detects calreticulin from canine, hamster, human, mouse and rat tissues. PA1-902A has been successfully used in Western blot, immunocytochemistry and immunofluorescene procedures. By Western blot, this antibody detects an ~60-66 kDa protein representing calreticulin. PA1-902A immunizing peptide corresponds to amino acid residues 24-43 from mouse calreticulin. PA1-902A immunizing peptide (Cat. # PEP-018) is available for use in neutralization and control experiments.

Calr or Calreticulin is a calcium-binding chaperone that promotes folding, oligomeric assembly and quality control in the endoplasmic reticulum (ER) via the calreticulin/calnexin cycle. Calr, a lectin, interacts transiently with almost all of the monoglucosylated glycoproteins that are synthesized in the ER. Calr also interacts with with the DNA-binding domain of NR3C1 and mediates its nuclear export. It may also be involved in maternal gene expression regulation and oocyte maturation via the regulation of calcium homeostasis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Calreticulin
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1mg/mL BSA and 0.05% sodium azide
Gene CALR
Gene Accession No. P14211, P18418, P27797, P28490, Q8K3H7
Gene Alias Autoantigen RO; CABP3; CALBP; calcium-binding protein 3; CALR; calrectulin; calregulin; Calreticulin; calreticulin precursor; cC1qR; CRP55; CRT; CRTC; Endoplasmic reticulum resident protein 60; epididymis secretory sperm binding protein Li 99n; ERp60; FLJ26680; grp60; HACBP; HEL-S-99n; I79_008346; RO; Ro/SS-A Antigen; Sicca syndrome antigen A (autoantigen Ro; calreticulin); SSA
Gene Symbols CALR
Host Species Chicken
Immunogen Synthetic Peptide: K(24) E Q F L D G D A W T N R W V E S K H K(43).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100689096, 12317, 476694, 64202, 811
Target Species Human, Mouse, Rat, Canine, Hamster
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgY
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.