Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Calretinin Monoclonal Antibody (SP13)

Catalog No. p-7167898
Change view
Click to view available options
Quantity:
500 μL
1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA139562 1 mL
PIMA516328 500 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PIMA139562 Supplier Invitrogen™ Supplier No. MA139562
Only null left
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

This antibody is predicted to react with bovine, chicken and rat based on sequence homology. Heat-mediated antigen retrieval is recommended prior to staining, using a 10mM citrate buffer, pH 6.0, for 10 minutes followed by cooling at room temperature for 20 min. Following antigen retrieval, incubate samples with primary antibody for 30 min at room temperature. A suggested positive control is mesothelioma.

Calretinin is encoded by the CALB2 gene located on chromosome 16. It is a troponin C superfamily protein having six EF-hand structural motifs. The EF motif allows the protein to bind calcium, and function in calcium signaling, calcium buffering, and modulating neuronal excitability. Calretinin is expressed in the mesothelium, mast cells, some neural cells, hair follicles, and fat cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Calretinin
Applications Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone SP13
Concentration Conc. Not Determined
Conjugate Unconjugated
Formulation PBS with 1% BSA and 0.1% sodium azide; pH 7.20
Gene CALB2
Gene Accession No. P22676, P47728, Q08331
Gene Alias 29 kDa calbindin; 29kD calretinin; 29kDa calbindin; CAB29; CAL2; Calb2; calbindin 2; calbindin 2, (29kD, calretinin); calbindin 2, 29kDa; calbindin D29K; calretinin; CR; testicular secretory protein Li 8
Gene Symbols CALB2
Host Species Rabbit
Immunogen Recombinant full-length mouse calretinin protein.
Purification Method Protein A
Quantity 1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 117059, 12308, 794
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.